Printer Ribbons, M710 R6600 Series
Supplier: Brady
BRDY173188EA 76.22 EUR
BRDY173188
Printer Ribbons, M710 R6600 Series
Printer Ribbons
One-color ribbon for printing a single color onto a label.
  • R6600 series resin formulation offers excellent solvent and friction resistance when printed on the appropriate Brady® label material
  • Recommended for printing on heat shrinkable tubing materials from Brady®, including material numbers B-342, B-7641, B-7642, B-7646, and B-7647
  • Self-contained M710 series ribbon cartridges insert straight into the printer with ease
M710 Series snap-in print ribbons enable 5 second ribbon supply changes. No calibrating, no threading, no waste, no hassles, and the ink always faces the right direction.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.