VWR®, EPA Method 1633 Reference Standard, 20 µg/L in acetonitrile, CRM
Supplier: Avantor
VWR®
89089.010EA 0 EUR
89089.010
VWR®, EPA Method 1633 Reference Standard, 20 µg/L in acetonitrile, CRM
Standards Environmental Analysis Standards
USEPA Method 1633 is for analysis of PFAS in aqueous, solid, biosolids and tissue samples using LC-MS/MS technique. Our mixes are offered to cover the 40 native PFAS required by the method.
Contains:
Sulfluramid4151-50-220 µg/mL
Perfluorooctane sulfonamide754-91-620 µg/mL
N-Methylperfluoro-1-octanesulfonamide31506-32-820 µg/mL
N-ethylperfluoro-1-octanesulfonamidoacetic acid2991-50-620 µg/mL
N-methylperfluoro-1-octanesulfonamidoacetic acid2355-31-920 µg/mL
Certifikace: ISO 17034