TIG Welding Gauntlets
Supplier: Portwest
A521BRRL A521BRRXL A521BRRXXL
PRTWA521BRRLEA 13.6 EUR
PRTWA521BRRL PRTWA521BRRXL PRTWA521BRRXXL
TIG Welding Gauntlets
Gloves
The TIG Ultra Welding Gauntlet offers fantastic dexterity with a goat leather palm and excellent burn resistance due to the split leather backing. Designed for the professional TIG welder with aramid stitching for ultra durability.
  • EN12477 Type A welding protection
  • Premium quality leather welding gauntlet
  • The ultimate in dexterity for welders
  • Made from high quality durable goat leather
Ideal for welding and metal handling, these gloves feature reinforced Aramid stitching for maximum durability and burn protection.
Colour: Brown
Certifikace: EN ISO 21420: 2020 Dexterity 2, EN 388: 2016 + A1: 2018 3243X, EN 12477: 2001 Type A, EN 407: 2020 422X31. ANSI/ISEA 105: 2016 CUT Level A2. CE certified.