rubis® Tweezers, PTFE-Coated
Supplier: OUTILS RUBIS
2A-FT 3-SA-FT 2A-SA-FT 35ASAFT
OUTI2A-FTEA 38.1 EUR
OUTI2A-FT OUTI3-SA-FT OUTI2A-SA-FT OUTI35ASAFT
rubis® Tweezers, PTFE-Coated
Full PTFE-coated (Teflon) tweezers made from stainless steel.
  • Made from anti-magnetic and anti-acid stainless steel
  • Good heat and chemical resistance
  • Excellent for use in Biotech applications
PTFE coating (Teflon) applied on anti-acid/anti-magnetic stainless steel for a smooth and flawless performance. PTFE coating provides a secure grip, and prevents scratches and almost any substance from adhering to the surface of the tweezers.