Pura-Fit® Classic Foam Earplugs
Supplier: Moldex
Pura-Fit®
MOLD776001EA 109 EUR
MOLD776001
Pura-Fit® Classic Foam Earplugs
Earplugs Disposable Earplugs
The classic Pura-Fit® earplugs offer the optimal combination of stability and softness. They are easy to roll and, upon insertion, expand to fill the ear canal at exactly the right moment. There, they maintain a secure, sealed fit, without exerting pressure on the ear.
  • Noise reduction at high (H), medium (M) and low (L) frequencies
  • 36 dB (SNR) noise reduction
  • Soft foam for a high degree of wearing comfort
  • Also available in a dispenser and with neck cord
Shelf life from date of production: 4 years.