Protein and peptide molecular weight markers
Supplier: Cytiva
17-0445-01
17-0446-01
17-0445-01EA
405
EUR
17-0445-01
17-0446-01
Protein and peptide molecular weight markers
Electrophoresis Reagents
Molecular Weight Markers
For determination of molecular weight of uncharacterised protein samples with precision by comparison with carefully selected standards.
- Clear, sharp bands for easy measurement of migration distances
- Bands can be visualised by using stains such as Deep Purple™ total protein stain, Coomassie® brilliant blue, or silver staining
Informace pro objednávání: The peptide marker kit is supplied with six horse myoglobin peptides in one 2 mg vial. The LMW-SDS marker kit is supplied with six proteins in ten 575 µg vials. The HMW-SDS kit is supplied with five proteins in ten 175 µg vials. The HMW native marker kit is supplied with five proteins in ten 250 µg vials.