FITC-xtra ≥90%
Supplier: AAT BIOQUEST
135 136
AATB135EA 307 EUR
AATB135 AATB136
FITC-xtra ≥90%
FITC-xtra
Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.
  • pH-Independent Fluorescence
  • High Signal/Background Ratio
Upozornení: For research use only.