Resazurin sodium salt ≥90%
Supplier: AAT BIOQUEST INC
|
Warning
|
AATB15700EA
115
EUR
AATB15700
Resazurin sodium salt ≥90%
Resazurin sodium salt
Resazurin is a non-toxic, water-soluble and redox-sensitive dye that changes from its blue/non-fluorescent state to pink/highly-fluorescent upon reduction to resorufin by many biological processes.
Formula:
C₁₂H₆NNaO₄ MW: 251,17 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...