Cal Green™ 1, AM Indicator_calcium
Supplier: AAT BIOQUEST
Warning
20501 20502
AATB20501EA 230 EUR
AATB20501 AATB20502
Cal Green™ 1, AM Indicator_calcium
Calcium Green 1AM
Cal Green™ 1 (AAT Bioquest) is the same molecule to Calcium Green-1 (Invitrogen).
  • Cell permeable
  • Calcium-dependent fluorescence
Formula: C₅₉H₅₃Cl₂N₃O₂₆
MW: 1290.96 g/mol
CAS Number: 186501-28-0
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=accf2d02e2f0f350e532bc4781c9386557b1e467&vid=e4d9b52904aab8cb09c5bff4b7eec4380abf54e5&prid=web&clid=SAPDAM