BAPTA-AM ≥95%, Ultra Pure Grade
Supplier: AAT BIOQUEST INC
|
Warning
|
AATB21002EA
146
EUR
AATB21002
BAPTA-AM ≥95%, Ultra Pure Grade
BAPTA-AM
BAPTA AM is cell-permeable version of BAPTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid), a cell-permeable calcium chelator.
Formula:
C₃₄H₄₀N₂O₁₈ MW: 764,69 g/mol |
MDL Number:
MFCD00036696 CAS Number: 126150-97-8 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...