Fluo-8L™ AM
Supplier: AAT BIOQUEST
Warning
21096 21097
AATB21096EA 460 EUR
AATB21096 AATB21097
Fluo-8L™ AM
Fluo-8L™ AM
Calcium measurements are critical for numerous biological investigations.
  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Suitable for all fluorescence platforms
Upozornení: For research use only.
MW: 1078.95 g/mol
ADR: 5,II