5-Carboxy-X-Rhodamine, NHS Ester 90
Supplier: AAT BIOQUEST
Warning
Synonyms: 5-ROX, SE, 5-ROX N-succinimidyl ester
389 391
AATB389EA 307 EUR
AATB389 AATB391
5-Carboxy-X-Rhodamine, NHS Ester 90
5-Carboxy-X-rhodamine, NHS ester
Single isomers of ROX are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purifications that are often required in bioconjugations.
Formula: C₃₇H₃₃N₃O₇
MW: 631.685 g/mol
CAS Number: 209734-74-7
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c908005b27be2106e8fe61c92dbe95ee17a3d58&vid=1b408fa5390e09ab585c3e3b08272bbe1cad0dee&prid=web&clid=SAPDAM