iFluor® 488, Tyramide
Supplier: AAT BIOQUEST INC
AATB45100EA 442 EUR
AATB45100
iFluor® 488, Tyramide
iFluor®
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
  • Increased fluorescence signal
  • Suitable for low-abundance targets
  • Compatible with all major fluorescence platforms
Upozornení: For research use only.