FMOC-Glu(TQ3)-OH for biomolecular labeling
Supplier: AAT BIOQUEST INC
|
Warning
|
AATB5238EA
590
EUR
AATB5238
FMOC-Glu(TQ3)-OH for biomolecular labeling
FMOC-Glu(TQ3)-OH
FMOC-Glu(TQ3)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 3 (TQ3).
- FRET building block
- Multiple applications
ADR:
5,II |