Bromocresol purple ≥95%, Ultra Pure Grade

Supplier: AAT BIOQUEST INC
Warning

AATB92EA 442 EUR
AATB92
Bromocresol purple ≥95%, Ultra Pure Grade
Bromocresol purple

Bromocresol purple (BCP) or 5',5-dibromo-o-cresolsulfophthalein, is a pH indicator.

Formula: C₂₁H₁₆Br₂O₅S
MW: 540,23 g/mol
MDL Number: MFCD00011681
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR