Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit
Supplier: AAT Bioquest
AATB22325EA 683 EUR
AATB22325
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit
Assays Cellular Assays
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
  • red laser compatible
  • highly reliable
  • optimized for flow cytometry
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit uses XdU which is incorporated into cellular DNA during DNA synthesis.
After fixing cells, the incorporated XdU is labelled with iFluor® 647 MTA. The resulted iFluor® 647-labeled DNA formed in cells is visualized in Cy5 Channel. Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay