Native Casein (from Eukaryotic cells), TAMRA (5-Carboxytetramethylrhodamin)
Supplier: AAT Bioquest
AATB13441EA 295 EUR
AATB13441
Native Casein (from Eukaryotic cells), TAMRA (5-Carboxytetramethylrhodamin)
Proteins and Peptides
Casein is considered to be a generic substrate for a broad spectrum of proteases.
  • Excellent fluorescence intensity change
  • Signal directly proportional to protease activity
  • Bright signal
  • Ph-independent fluorescence
  • Suitable for low ph
Upozornení: For research use only.