Rhodium ≥99.8% (metals basis), wire, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
11576.BS 11576.BR 11576.BV 11576.CB
11576.BSEA 703 EUR
11576.BS 11576.BR 11576.BV 11576.CB
Rhodium ≥99.8% (metals basis), wire, Ø 0.5 mm (0.02 in)
Rhodium
Formula: Rh
MW: 102.9 g/mol
Boiling Pt: 3400…3700 °C (1013 hPa)
Melting Pt: 1960
Density: 12.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6
EINECS: 231-125-0
Merck Index: 12,08353