2,4-Pentanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,4-Dihydroxy pentane
M-Clarity™ quality level = MQ100
156019-10G 156019-50G
SIAL156019-10GEA 92.7 EUR
SIAL156019-10G SIAL156019-50G
2,4-Pentanediol, Sigma-Aldrich®
2,4-Pentanediol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 201
Melting Pt: ∼53 °C
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 101
Storage Temperature: Ambient
MDL Number: MFCD00004549
CAS Number: 625-69-4
EINECS: 210-907-5