Ammonium chloride-[D4], Sigma-Aldrich®

Supplier: Merck

Synonyms: Ammonium-d4 chloride

M-Clarity™ quality level = MQ200

175676-10G 175676-5G
SIAL175676-10GEA 251 EUR
SIAL175676-10G SIAL175676-5G
Ammonium chloride-[D4], Sigma-Aldrich®
Ammonium chloride-[D4]
Formula: ND₄Cl
MW: 57.46 g/mol
Melting Pt: 335 °C
Density: 1.52 g/cm³ (20 °C)
CAS Number: 12015-14-4
EINECS: 234-607-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR