Native Horseradish Horseradish peroxidase (from Serum), 3-Amino-2-oxazolidine (AOZ)
Supplier: AAT Bioquest
AATB50006EA 739 EUR
AATB50006
Native Horseradish Horseradish peroxidase (from Serum), 3-Amino-2-oxazolidine (AOZ)
Proteins and Peptides
3-Amino-2-oxazolidine (AOZ)-HRP conjugate is used for developing an ELISA assay for quantifying 3-Amino-2-oxazolidine (AOZ).
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Upozornení: For research use only.