Fluo-8H™ AM
Supplier: AAT BIOQUEST
Warning
21090 21091
AATB21090EA 460 EUR
AATB21090 AATB21091
Fluo-8H™ AM
Fluo-8H™ AM
Calcium measurements are critical for numerous biological investigations.
  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Suitable for all fluorescence platforms
Upozornení: For research use only.
MW: 1074.98 g/mol
ADR: 5,II