Trypan 0.1 M, Trypan Red Plus™ for dead cell labeling
Supplier: AAT BIOQUEST
Warning
2456 2457
AATB2456EA 307 EUR
AATB2456 AATB2457
Trypan 0.1 M, Trypan Red Plus™ for dead cell labeling
Trypan
Trypan red Plus™ is similar to Trypan Blue in cell permeability.
  • Minimal cytotoxicity
  • High purity
MW: 600 g/mol
ADR: 5 II