Gibberellic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
48880-250MG-F 48880-1G-F 48880-5G-F
SIAL48880-250MG-FEA 43.7 EUR
SIAL48880-250MG-F SIAL48880-1G-F SIAL48880-5G-F
Gibberellic acid, Sigma-Aldrich®
Gibberellic acid
Formula: C₁₉H₂₂O₆
MW: 346.38 g/mol
Melting Pt: 234
Density: 1.386 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00079329
CAS Number: 77-06-5
EINECS: 201-001-0
Merck Index: 13,04430