Polyethylene glycol 8,000, Sigma-Aldrich®
Supplier: Merck
Synonyms: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000
MQ200
202452-250G 202452-500G
SIAL202452-250GEA 43.8 EUR
SIAL202452-250G SIAL202452-500G
Polyethylene glycol 8,000, Sigma-Aldrich®
Polyethylene glycol 8,000
Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Boiling Pt: 250
Density: 1.125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729