(±)-3,5-Dimethyl-1-hexyn-3-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms: (±)-2,4-Dimethyl-5-hexyn-4-ol, 3,5-Dimethylhex-1-yn-3-ol, 3,5-Dimethyl-1-hexyn-3-ol
M-Clarity™ quality level = MQ200
278394-100ML 278394-500ML
SIAL278394-100MLEA 48.6 EUR
SIAL278394-100ML SIAL278394-500ML
(±)-3,5-Dimethyl-1-hexyn-3-ol, Sigma-Aldrich®
(±)-3,5-Dimethyl-1-hexyn-3-ol
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 150 °C (1013 hPa)
Density: 0.859 g/cm³ (12 °C)
MDL Number: MFCD00008941
CAS Number: 107-54-0
EINECS: 203-500-9