1-Iodoperfluorbutane tech
Supplier: Apollo Scientific
Warning
Synonyms: Perfluorobutyl iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, Nonafluoro-1-iodobutane
PC99446-25G PC99446-100G PC99446-500G
APOSPC99446-25GEA 23.2 EUR
APOSPC99446-25G APOSPC99446-100G APOSPC99446-500G
1-Iodoperfluorbutane tech
1-Iodoperfluorbutane
Formula: C₄F₉I
MW: 345.93 g/mol
Boiling Pt: 64…66 °C
Melting Pt: –88 °C
Density: 2.01
Flash Pt: >115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001062
CAS Number: 423-39-2
UN: 3082
ADR: 9+EHS,III