1-(4-Bromo-3-nitrophenyl)-1-butanone 98%
Supplier: Apollo Scientific
Warning
OR450141-250MG OR450141-1G OR450141-5G
APOSOR450141-250MGEA 241 EUR
APOSOR450141-250MG APOSOR450141-1G APOSOR450141-5G
1-(4-Bromo-3-nitrophenyl)-1-butanone 98%
1-(4-Bromo-3-nitrophenyl)-1-butanone
Formula: C₁₀H₁₀BrNO₃
MW: 272.1 g/mol
MDL Number: MFCD34474617
CAS Number: 91715-78-5