4-(Dihydroxyborophenyl)acetylene 98%
Supplier: Apollo Scientific
Warning
Synonyms: B-(4-Ethynylphenyl)boronic Acid, (4-Ethynylphenyl)boronic Acid, 4-Ethynylbenzeneboronic Acid
OR361136-250MG OR361136-1G OR361136-5G
APOSOR361136-250MGEA 68.3 EUR
APOSOR361136-250MG APOSOR361136-1G APOSOR361136-5G
4-(Dihydroxyborophenyl)acetylene 98%
4-(Dihydroxyborophenyl)acetylene
Formula: C₈H₇BO₂
MW: 145.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08689553
CAS Number: 263368-72-5