4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) (free radical) tech
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Amino-1-oxy-2,2,6,6-tetramethylpiperidine
OR019107-5G OR019107-25G OR019107-100G
APOSOR019107-5GEA 23.2 EUR
APOSOR019107-5G APOSOR019107-25G APOSOR019107-100G
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) (free radical) tech
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-Amino-TEMPO, free radical)
Formula: C₉H₁₉N₂O
MW: 171.26 g/mol
Boiling Pt: 98/3.75 mm °C
Melting Pt: 33 - 39 °C
Density: 0.896
Storage Temperature: Refrigerator
MDL Number: MFCD00006479
CAS Number: 14691-88-4