1-Methylnaphthalene 96%
Supplier: Apollo Scientific
Warning
Synonyms: α-Methylnaphthalene
OR019113-25G OR019113-100G OR019113-500G
APOSOR019113-25GEA 58 EUR
APOSOR019113-25G APOSOR019113-100G APOSOR019113-500G
1-Methylnaphthalene 96%
1-Methylnaphthalene
Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 240…243 °C
Melting Pt: –22 °C
Density: 1.001
Flash Pt: 82 °C
MDL Number: MFCD00004034
CAS Number: 90-12-0
UN: 3082