(S)-tert-Butyl 3-aminoazepane-1-carboxylate
Supplier: Apollo Scientific
Danger
OR918689-250MG OR918689-1G
APOSOR918689-250MGEA 108 EUR
APOSOR918689-250MG APOSOR918689-1G
(S)-tert-Butyl 3-aminoazepane-1-carboxylate
(S)-tert-Butyl 3-aminoazepane-1-carboxylate
Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
MDL Number: MFCD11041390
CAS Number: 625471-04-7