1,2,4-Triazol-5-one 98%
Supplier: Apollo Scientific
Warning
OR931823-1G OR931823-5G OR931823-25G
APOSOR931823-1GEA 24.8 EUR
APOSOR931823-1G APOSOR931823-5G APOSOR931823-25G
1,2,4-Triazol-5-one 98%
1,2,4-Triazol-5-one
Formula: C₂HN₃O
MW: 83.05 g/mol
MDL Number: MFCD01446029
CAS Number: 42131-33-9