2-(Difluoromethylsulphanyl)isoindole-1,3-dione 98%
Supplier: Apollo Scientific
Warning
PC48672-250MG PC48672-1G
APOSPC48672-250MGEA 219 EUR
APOSPC48672-250MG APOSPC48672-1G
2-(Difluoromethylsulphanyl)isoindole-1,3-dione 98%
2-(Difluoromethylsulphanyl)isoindole-1,3-dione
Formula: C₉H₅F₂NO₂S
MW: 229.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32066858
CAS Number: 1805773-37-8