1-(tert-Butyldimethylsilyloxy)-1-methoxyethene
Supplier: Apollo Scientific
Danger
Synonyms: tert-Butyl((1-methoxyvinyl)oxy)dimethylsilane, tert-Butyl[(1-methoxyvinyl)oxy]dimethylsilane
OR923452-1G OR923452-5G
APOSOR923452-1GEA 58 EUR
APOSOR923452-1G APOSOR923452-5G
1-(tert-Butyldimethylsilyloxy)-1-methoxyethene
1-(tert-Butyldimethylsilyloxy)-1-methoxyethene
Formula: C₉H₂₀O₂Si
MW: 188.34 g/mol
Boiling Pt: 62/9 mm °C
Density: 0.863
Flash Pt: 52 °C
MDL Number: MFCD02683524
CAS Number: 77086-38-5
UN: 1993
ADR: 3 III