Ethyl 4-bromo-2-chlorobenzoate 98%
Supplier: Apollo Scientific
Warning
OR956775-1G OR956775-5G
APOSOR956775-1GEA 238 EUR
APOSOR956775-1G APOSOR956775-5G
Ethyl 4-bromo-2-chlorobenzoate 98%
Ethyl 4-bromo-2-chlorobenzoate
Formula: C₉H₈BrClO₂
MW: 263.51 g/mol
MDL Number: MFCD10000926
CAS Number: 76008-74-7