1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbaldehyde 95%
Supplier: Apollo Scientific
Warning
PC408443-250MG PC408443-1G PC408443-5G
APOSPC408443-250MGEA 219 EUR
APOSPC408443-250MG APOSPC408443-1G APOSPC408443-5G
1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbaldehyde 95%
1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbaldehyde
Formula: C₆H₅F₃N₂O
MW: 178.11 g/mol
MDL Number: MFCD28403393
CAS Number: 1557546-42-5