3-Bromo-5-chloro-2-iodobenzaldehyde
Supplier: Apollo Scientific
Warning
OR61186-1G OR61186-5G
APOSOR61186-1GEA 574 EUR
APOSOR61186-1G APOSOR61186-5G
3-Bromo-5-chloro-2-iodobenzaldehyde
3-Bromo-5-chloro-2-iodobenzaldehyde
Formula: C₇H₃BrClIO
MW: 345.36 g/mol
MDL Number: MFCD30834285
CAS Number: 2091701-23-2