Heptafluoro-1-(1-iodo-1,2,2,2-tetrafluoroethoxy)propane
Supplier: Apollo Scientific
Warning
PC4514E-250MG PC4514E-1G
APOSPC4514E-250MGEA 29 EUR
APOSPC4514E-250MG APOSPC4514E-1G
Heptafluoro-1-(1-iodo-1,2,2,2-tetrafluoroethoxy)propane
Heptafluoro-1-(1-iodo-1,2,2,2-tetrafluoroethoxy)propane
Formula: C₅F₁₁IO
MW: 411.94 g/mol
Boiling Pt: 84…85 °C
Density: 1.9859
MDL Number: MFCD00155845
CAS Number: 107432-46-2