7-Methyl-4-indanol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
7-Methyl-2,3-dihydro-1H-inden-4-ol, 7-Methylindan-4-ol
OR21818-1G
OR21818-5G
OR21818-10G
APOSOR21818-1GEA
116
EUR
APOSOR21818-1G
APOSOR21818-5G
APOSOR21818-10G
7-Methyl-4-indanol
7-Methyl-4-indanol
Formula:
C₁₀H₁₂O MW: 148.2 g/mol |
MDL Number:
MFCD00173675 CAS Number: 16400-13-8 |