2-Pyrrolidone
Supplier: Apollo Scientific
Warning
Synonyms: Pyrrolidin-2-one
OR922249-5G OR922249-25G OR922249-100G
APOSOR922249-5GEA 21.9 EUR
APOSOR922249-5G APOSOR922249-25G APOSOR922249-100G
2-Pyrrolidone
2-Pyrrolidone
Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 245 °C
Melting Pt: 23…25 °C
Flash Pt: 113 °C
MDL Number: MFCD00005270
CAS Number: 616-45-5
Merck Index: 12,08200