1-Chloroisoquinoline tech
Supplier: Apollo Scientific
Warning
OR61190-5G OR61190-25G
APOSOR61190-5GEA 21.9 EUR
APOSOR61190-5G APOSOR61190-25G
1-Chloroisoquinoline tech
1-Chloroisoquinoline
Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 274…275/768 mm °C
Melting Pt: 31…36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00024134
CAS Number: 19493-44-8