2-Acetamido-4-fluorobenzoic acid
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Acetylamino)-4-fluorobenzoic acid, N-Acetyl-4-fluoroanthranilic acid
PC0673-100MG PC0673-1G PC0673-5G
APOSPC0673-100MGEA 21.9 EUR
APOSPC0673-100MG APOSPC0673-1G APOSPC0673-5G
2-Acetamido-4-fluorobenzoic acid
2-Acetamido-4-fluorobenzoic acid
Formula: C₉H₈FNO₃
MW: 197.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00665891
CAS Number: 394-27-4