3-Bromo-2-iodobenzotrifluoride 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-2-iodo-alpha,alpha,alpha-trifluorotoluene, 1-Bromo-2-iodo-3-(trifluoromethyl)benzene
PC48519-250MG PC48519-1G
APOSPC48519-250MGEA 192 EUR
APOSPC48519-250MG APOSPC48519-1G
3-Bromo-2-iodobenzotrifluoride 95%
3-Bromo-2-iodobenzotrifluoride
Formula: C₇H₃BrF₃I
MW: 350.9 g/mol
Boiling Pt: 85/1.5 mm °C
Melting Pt: 34…36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13194205
CAS Number: 85977-23-7