(R)-(-)-4-Phenyl-2-butanol 98%
Supplier: Apollo Scientific
Warning
Synonyms: (R)-(-)-4-Phenylbutan-2-ol, (R)-4-Phenylbutan-2-ol
OR304056-1G OR304056-5G
APOSOR304056-1GEA 55.1 EUR
APOSOR304056-1G APOSOR304056-5G
(R)-(-)-4-Phenyl-2-butanol 98%
(R)-(-)-4-Phenyl-2-butanol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 239 °C
Density: 0.976
Flash Pt: >110 °C
MDL Number: MFCD03093081
CAS Number: 39516-03-5