2,4-Dihydroxybenzoic acid 97%
Supplier: Apollo Scientific
Danger
Synonyms: beta-Resorcylic acid
OR4893-500G OR4893-100G OR4893-1KG
APOSOR4893-500GEA 72.5 EUR
APOSOR4893-500G APOSOR4893-100G APOSOR4893-1KG
2,4-Dihydroxybenzoic acid 97%
2,4-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 390
Melting Pt: 208…218 °C
MDL Number: MFCD00002451
CAS Number: 89-86-1
Merck Index: 12,08325