1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole, 1-Methyl-1h-indazol-3-ylboronic acid pinacol ester
OR908923-1G OR908923-5G
APOSOR908923-1GEA 354 EUR
APOSOR908923-1G APOSOR908923-5G
1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole 98%
1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
Formula: C₁₄H₁₉BN₂O₂
MW: 258.12 g/mol
Storage Temperature: Freezer
MDL Number: MFCD13182067
CAS Number: 1627722-97-7