1-Bromo-4,5-difluoro-2-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-4,5-difluoronitrobenzene
PC502293-5G PC502293-10G
APOSPC502293-5GEA 21.9 EUR
APOSPC502293-5G APOSPC502293-10G
1-Bromo-4,5-difluoro-2-nitrobenzene 98%
1-Bromo-4,5-difluoro-2-nitrobenzene
Formula: C₆H₂BrF₂NO₂
MW: 237.98 g/mol
Boiling Pt: 225.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06808551
CAS Number: 321-17-5