2,6-Difluoro-3-(trifluoromethoxy)benzaldehyde 95%
Supplier: Apollo Scientific
Warning
PC502464-250MG PC502464-1G PC502464-5G
APOSPC502464-250MGEA 190 EUR
APOSPC502464-250MG APOSPC502464-1G APOSPC502464-5G
2,6-Difluoro-3-(trifluoromethoxy)benzaldehyde 95%
2,6-Difluoro-3-(trifluoromethoxy)benzaldehyde
Formula: C₈H₃F₅O₂
MW: 226.1 g/mol
MDL Number: MFCD31561158
CAS Number: 2244087-04-3