7-Bromo-3-(difluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine 95%
Supplier: Apollo Scientific
Warning
PC502948-250MG PC502948-1G
APOSPC502948-250MGEA 400 EUR
APOSPC502948-250MG APOSPC502948-1G
7-Bromo-3-(difluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine 95%
7-Bromo-3-(difluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formula: C₇H₄BrF₂N₃
MW: 248.03 g/mol
MDL Number: MFCD31922795
CAS Number: 2331325-26-7